missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIAE Monoclonal Antibody (03)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA529569
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SIAE/sialate O-acetylesterase.
This gene encodes an enzyme which removes 9-O-acetylation modifications from sialic acids. Mutations in this gene are associated with susceptibility to autoimmune disease 6. Multiple transcript variants encoding different isoforms, found either in the cytosol or in the lysosome, have been found for this gene.
Specifications
| SIAE | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9HAT2 | |
| SIAE | |
| Recombinant Human sialate O-acetylesterase Protein (Met1-Lys523). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 03 | |
| Unconjugated | |
| SIAE | |
| AIS6; clone 165; CSEC; CSE-C; cytosolic sialic acid 9-O-acetylesterase homolog; H-Lse; LSE; RGD1310431; SIAE; Sialate O-acetylesterase; Sialate O-acetylesterase large subunit; Sialate O-acetylesterase small subunit; sialic acid acetylesterase; sialic acid-specific 9-O-acetylesterase; sialic acid-specific acetylesterase II; yolk sac gene 2; yolk sac protein 2; YSG2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 54414 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction