missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SI-CLP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558761
This item is not returnable.
View return policy
Description
Immunogen sequence: KTLLEKSQFS DKPVQDRGLV VTDLKAESVV LEHRSYCSAK ARDRHFAGDV LGYVTPWNSH GYDVTKVFGS KFTQISPV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.
SI-CLP detects human Stabilin-interacting chitinase like protein (SI-CLP), a novel member of the Glyco-18-domain containing protein family. SI-CLP is a secretory protein which is up-regulated in response to interleukin-4 and dexamethasone in activated macrophages. SI-CLP is also expressed by T- and B-lymphocytes, and by cells of monocytic and epithelial origin. SI-CLP is the ligand for stabilin-1, an intracellular sorting receptor which sorts SI-CLP into late endosomes and secretory lysosomes in activated macrophages.
Specifications
| SI-CLP | |
| Polyclonal | |
| Unconjugated | |
| CHID1 | |
| 3110023E09Rik; AI841869; CHID1; chitinase domain containing 1; chitinase domain-containing protein 1; FLJ42707; GL008; MGC3234; PSEC0104; SB139; SICLP; SI-CLP; stabilin-1 interacting chitinase-like protein; stabilin-1-interacting chitinase-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 66005 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BWS9 | |
| CHID1 | |
| Recombinant protein corresponding to Human SI-CLP. Recombinant protein control fragment (Product #RP-97509). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction