missing translation for 'onlineSavingsMsg'
Learn More

Shvo's Catalyst, TRC

Product Code. 30419742
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30419742 500 mg
30415828 100 mg
30415827 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30419742 Supplier Toronto Research Chemicals Supplier No. TRCS282400500MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104439-77-2
Molecular Formula C62H42O6Ru2
Molecular Weight (g/mol) 1085.13
Synonym Tetracarbonyl-μ-hydro[(1,2,3,4,5-η)-1-hydroxy-2,3,4,5-tetraphenyl-2,4-cyclopentadien-1-yl][(2,3,4,5-η)-2,3,4,5-tetraphenyl-2,4-cyclopentadien-1-one]di-ruthenium Coordination Compound, 2,3,4,5-Tetraphenyl-1,3-cyclopentadien-1-ol Ruthenium Complex, 2,3,4,5-Tetraphenyl-2,4-cyclopentadien-1-one Ruthenium Complex, Tetracarbonyl-μ-hydro[(1,2,3,4,5-η)-1-hydroxylato-2,3,4,5-tetraphenyl-2,4-cyclopentadien-1-yl][(1,2,3,4,5-η)-1-hydroxy-2,3,4,5-tetraphenyl-2,4-cyclopentadien-1-yl]diruthenium, 1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium(II), Shvo Catalyst
SMILES C1=CC=C(C=C1)C2=C(C3=CC=CC=C3)C(=C([C]2C4=CC=CC=C4)O)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C(C3=CC=CC=C3)C(=O)C(=C2C4=CC=CC=C4)C5=CC=CC=C5.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Ru].[Ru]

Specifications

Quantity 500 mg
Chemical Name or Material Shvo's Catalyst
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.