Learn More
SymLine™ Angled Shut-off Adapter
Product Code. 13508840
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Closure Size:
GL25
missing translation for 'unitSize'
Each
Product Code. Closure Size unitSize
13508840 GL25 Each
1 missing translation for 'options'
Product Code. 13508840 missing translation for 'mfr' SymLine™ missing translation for 'supplierNo' 106475
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
  • Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
  • All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Closure Size GL25
Closure Color Black
Description 1 x GL25 (f) to 1 x GL25 (m)
Color Black
Liner Material HDPE-EC
Material HDPE-EC
Product Type Adapter