missing translation for 'onlineSavingsMsg'
Learn More

SHROOM3 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18407445 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407445 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407445 Supplier Novus Biologicals Supplier No. NBP281883UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SHROOM3 Polyclonal antibody specifically detects SHROOM3 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHROOM3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias APXL3, MSTP013, SHRM, ShrmL, SHROOM3 shroom family member 3
Host Species Rabbit
Immunogen Rabbit polyclonal SHROOM3 antibody was raised against an 18 amino acid peptide near the center of human SHROOM3. The immunogen is located within amino acids 850 - 900 of SHROOM3. Amino Acid Squence: CQLSGGASDSGRGPQRPDA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57619
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.