missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SHROOM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562449
This item is not returnable.
View return policy
Description
Immunogen sequence: PPLLIQDEDS TRIERVMDNN TTVKMVPIKI VHSESQPEKE SRQSLACPAE PPALPHGLEK DQIKTLSTSE QFYS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.
The protein encoded by this gene shares significant similarities with the apical protein from Xenopus laevis which is implicated in amiloride-sensitive sodium channel activity. This gene is a strong candidate gene for ocular albinism type 1 syndrome.
Specifications
| SHROOM2 | |
| Polyclonal | |
| Unconjugated | |
| SHROOM2 | |
| 4832440C16; Ab2-404; apical-like protein; APX homolog of Xenopus; Apxl; C630003H05Rik; HSAPXL; liver regeneration-related protein LRRG167; protein Apxl; Protein Shroom2; Shrm2; shroom family member 2; SHROOM2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 357 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13796 | |
| SHROOM2 | |
| Recombinant protein corresponding to Human SHROOM2. Recombinant protein control fragment (Product #RP-95447). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction