missing translation for 'onlineSavingsMsg'
Learn More

Shrew-1/AJAP1 Antibody [DyLight 550], Novus Biologicals™

Product Code. 30732312 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732312 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732312 Supplier Novus Biologicals Supplier No. AF7970L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Shrew-1/AJAP1 Polyclonal antibody specifically detects Shrew-1/AJAP1 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Shrew-1/AJAP1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias adherens junction associated protein 1, adherens junction-associated protein 1, adherens junctions associated protein 1, membrane protein shrew-1, MOT8, RP3-426F10.1, SHREW1, SHREW-1, transmembrane protein SHREW1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Shrew-1/AJAP1, His157-His283, Accession # A2ALI5
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55966
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.