missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SHQ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572672
This item is not returnable.
View return policy
Description
SHQ1 Polyclonal Antibody for Western Blot
SHQ1 is the human homolog of yeast Shq1p, a protein that acts as an accessory factor in the assembly of H/ACA ribonucleoproteins. SHQ1 has been shown to bind the major H/ACA core protein and the pseudouridine synthase NAP57, but precedes the assembly role of NAF1 and of the other H/ACA core proteins. SHQ1 assists in the assembly of H/ACA-box ribonucleoproteins that function in the processing of ribosomal RNAs, modification of spliceosomal small nuclear RNAs, and stabilization of telomerase.
Specifications
| SHQ1 | |
| Polyclonal | |
| Unconjugated | |
| SHQ1 | |
| 2810403P18Rik; FLJ10539; gene associated with retinoid and interferon-induced mortality 1; GRIM-1; hypothetical protein LOC507486; protein SHQ1 homolog; RGD1310610; Shq1; SHQ1 homolog; SHQ1 homolog (S. cerevisiae); SHQ1, H/ACA ribonucleoprotein assembly factor; Shq1p; zgc:158255 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 567601 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.7 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| A1L1R0 | |
| SHQ1 | |
| KLH conjugated synthetic peptide between 573-605 amino acids from the C-terminal region of DANRE shq1. | |
| 400 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction