missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHQ1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-81793CL1
This item is not returnable.
View return policy
Description
SHQ1 Polyclonal antibody specifically detects SHQ1 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| SHQ1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 55164 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| DKFZp686H07226, FLJ10539, protein SHQ1 homolog, SHQ1 homolog (S. cerevisiae), Shq1p | |
| SHQ1 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human SHQ1. The immunogen is located within amino acids 80 - 130 of SHQ1. Amino Acid Squence: LAPRKSRTAKPLVEEIG | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction