missing translation for 'onlineSavingsMsg'
Learn More

SHQ1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18415837 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415837 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415837 Supplier Novus Biologicals Supplier No. NBP281793AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SHQ1 Polyclonal antibody specifically detects SHQ1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHQ1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias DKFZp686H07226, FLJ10539, protein SHQ1 homolog, SHQ1 homolog (S. cerevisiae), Shq1p
Host Species Rabbit
Immunogen SHQ1 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human SHQ1. The immunogen is located within amino acids 80 - 130 of SHQ1. Amino Acid Squence: LAPRKSRTAKPLVEEIG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55164
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.