missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SHPRH Recombinant Protein Antigen

Product Code. 18323303
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323303 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323303 Supplier Novus Biologicals™ Supplier No. NBP317931PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 257218
Molecular Weight (g/mol) M.W. Theoretical: 30 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias 2610103K11Rik, bA545I5.2, E3 ubiquitin-protein ligase SHPRH, EC 3.6.4.-, EC 6.3.2.-, FLJ27258, FLJ37625, FLJ90837, histone-linker, PHD and RING finger domain-containing helicase, KIAA2023FLJ45012, MGC134886, SNF2 histone linker PHD RING helicase
Gene Symbol SHPRH
Research Category DNA Repair
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein SHPRH
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.