missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHPRH Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21955-1-AP-20UL
This item is not returnable.
View return policy
Description
A novel gene containing SNF2, PHD-finger, RING-finger, helicase, and linker histone domains termed SHPRH, is a possible candidate for a tumor suppressor gene. These domains are characteristic of several DNA repair proteins, transcription factors, and helicases. Mapping of this gene to 6q24, a region reported to contain a tumor suppressor locus suggests a role in tumor suppression.Specifications
| SHPRH | |
| Polyclonal | |
| Unconjugated | |
| SHPRH | |
| KIAA2023, SHPRH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 257218, 268281 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q149N8, Q7TPQ3 | |
| SHPRH | |
| SHPRH Fusion Protein Ag16635 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction