missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SHPRH Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550788
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SHPRH protein.
A novel gene containing SNF2, PHD-finger, RING-finger, helicase, and linker histone domains termed SHPRH, is a possible candidate for a tumor suppressor gene. These domains are characteristic of several DNA repair proteins, transcription factors, and helicases. Mapping of this gene to 6q24, a region reported to contain a tumor suppressor locus suggests a role in tumor suppression.
Specifications
| SHPRH | |
| Polyclonal | |
| Unconjugated | |
| SHPRH | |
| 2610103K11Rik; AA450458; AU024614; bA545I5.2; BC006883; D230017O13Rik; E130018M05; E3 ubiquitin-protein ligase SHPRH; KIAA2023; RING-type E3 ubiquitin transferase SHPRH; Shprh; SNF2 histone linker PHD RING helicase; SNF2 histone linker PHD RING helicase, E3 ubiquitin protein ligase; SNF2, histone-linker, PHD and RING finger domain-containing helicase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 257218 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.4 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q149N8 | |
| SHPRH | |
| Fusion protein corresponding to residues near the C terminal of human SNF2 histone linker PHD RING helicase, E3 ubiquitin protein ligase. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction