missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHPK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12441-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene has weak homology to several carbohydrate kinases, a class of proteins involved in the phosphorylation of sugars as they enter a cell, inhibiting return across the cell membrane. Sequence variation between this novel gene and known carbohydrate kinases suggests the possibility of a different substrate, cofactor or changes in kinetic properties distinguishing it from other carbohydrate kinases. The gene resides in a region commonly deleted in cystinosis patients, suggesting a role as a modifier for the cystinosis phenotype. The genomic region is also rich in Alu repetitive sequences, frequently involved in chromosomal rearrangements.Specifications
| SHPK | |
| Polyclonal | |
| Unconjugated | |
| SHPK | |
| CARKL, sedoheptulokinase, SHK, SHPK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23729, 287479, 74637 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9D5J6, Q9UHJ6 | |
| Shpk | |
| SHPK Fusion Protein Ag3150 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction