missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHP/NR0B2/Nuclear Receptor SHP Antibody (OTI7B5), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74172G
This item is not returnable.
View return policy
Description
SHP/NR0B2/Nuclear Receptor SHP Monoclonal specifically detects SHP/NR0B2/Nuclear Receptor SHP in Human samples. It is validated for Western Blot.
Specifications
| SHP/NR0B2/Nuclear Receptor SHP | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NR0B2 | |
| Full length recombinant protein of human SHP/NR0B2/Nuclear Receptor SHP (NP_068804) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot | |
| OTI7B5 | |
| Western Blot | |
| FLJ17090, nuclear receptor subfamily 0 group B member 2, nuclear receptor subfamily 0, group B, member 2, Orphan nuclear receptor SHP, SHPSHP1, Small heterodimer partner | |
| Mouse | |
| Affinity Purified | |
| GPCR | |
| 8431.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction