missing translation for 'onlineSavingsMsg'
Learn More

SHP-2/PTPN11, p Tyr542 Antibody (2E2), Novus Biologicals™

Product Code. p-200022295 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30027700 50 μL 50µL
30036097 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30027700 Supplier Novus Biologicals Supplier No. NBP32639750ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SHP-2/PTPN11 Monoclonal antibody specifically detects SHP-2/PTPN11 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHP-2/PTPN11
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 2E2
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11
Host Species Rabbit
Immunogen A synthesized peptide derived from Human SHP-2/PTPN11 [p Tyr542] [UniProt Q06124]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Lipid and Metabolism, Phospho Specific, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 5781
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.