missing translation for 'onlineSavingsMsg'
Learn More

SHP-2/PTPN11 Antibody (OTI3F2), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18405067 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405067 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405067 Supplier Novus Biologicals Supplier No. NBP274173MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SHP-2/PTPN11 Monoclonal antibody specifically detects SHP-2/PTPN11 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHP-2/PTPN11
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3F2
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11
Host Species Mouse
Immunogen Full length human recombinant protein of human PTPN11(NP_002825) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Lipid and Metabolism, Phospho Specific, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 5781
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.