missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHP-1 Recombinant Rabbit Monoclonal Antibody (3H20L13)
SHP-1 Recombinant Rabbit Monoclonal Antibody (3H20L13)
Brand: Invitrogen 703568
This item is not returnable.
View return policy
Description
SHP-1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Specifications
| SHP-1 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P29350 | |
| PTPN6 | |
| Protein corresponding to human SHP-1 [aa243-aa541]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry, Western Blot | |
| 3H20L13 | |
| Unconjugated | |
| PTPN6 | |
| 70Z-SHP; EC 3.1.3.48; hcp; Hcph; hematopoietic cell phosphatase; hematopoietic cell protein-tyrosine phosphatase; HPTP1C; me; motheaten; protein tyrosine phosphatase, non-receptor type 6; protein-tyrosine phosphatase 1C; Protein-tyrosine phosphatase SHP-1; PTN6; Ptp1C; PTP-1C; Ptph6; PTPN6; PTPTY-42; SH2 phosphatase 1; SHP1; Shp-1; SHP-1L; SH-PTP1; Tyrosine-protein phosphatase non-receptor type 6 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5777 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction