missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHP-1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-60553
This item is not returnable.
View return policy
Description
SHP-1 Polyclonal specifically detects SHP-1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| SHP-1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| HCP, HCPH, HPTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_536858.1and NP_002822.2) | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.2 - 0.5 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| NP_536858.1 | |
| PTPN6 | |
| Peptide with sequence C-HTKNKREEKVKKQ corresponding to internal region (near C-Terminus) according to NP_536858.1, NP_002822.2. | |
| 0.1 mg | |
| Apoptosis, Protein Phosphatase | |
| 5777 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction