missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SHOX2 Polyclonal specifically detects SHOX2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | SHOX2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 - 1:5000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | Homeobox protein Og12X, OG12SHOX homologous gene on chromosome 3, OG12Xshort stature homeobox protein 2, short stature homeobox 2, SHOTPaired-related homeobox protein SHOT |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 96-355 of human SHOX2 (NP_003021.3). ELDMGAAERSREPGSPRLTEGRRKPTKAEVQATLLLPGEAFRFLVSPELKDRKEDAKGMEDEGQTKIKQRRSRTNFTLEQLNELERLFDETHYPDAFMREELSQRLGLSEARVQVWFQNRRAKCRKQENQLHKGVLIGAASQFEACRVAPYVNVGALRMPFQQDSHCNVTPLSFQVQAQLQLDSAVAHAHHHLHPHLAAHAPYMMFPAPPFGLPLATLAADSASAASVVAAAAAAKTTSKNSSIADLRLKAKKHAAALGL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?