missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOX Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | SHOX |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18348154
|
Novus Biologicals
NBP3-15753-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18365054
|
Novus Biologicals
NBP3-15753-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHOX Polyclonal antibody specifically detects SHOX in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SHOX | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| GCFXX-linked, PHOGShort stature homeobox-containing protein, Pseudoautosomal homeobox-containing osteogenic protein, short stature homeobox, short stature homeobox protein, SHOXY | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-110 of human SHOX (NP_000442.1). | |
| Primary | |
| Store at 4°C. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 6473 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title