missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SHOX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566351
This item is not returnable.
View return policy
Description
Immunogen sequence: GTSDSSLQDIT EGGGHCPVHL FKDHVDNDKE KLKEFGTARV AEGIYECKEK REDVK Highest antigen sequence identity to the following orthologs - mouse 27%, rat 30%.
Homeodomain proteins (HP) are transcriptional regulators that coordinate the expression of genes involved in development, differentiation and cellular transformation. HPs are characterized by a conserved domain of 60 amino acid residues that recognize and bind a site in the regulatory region of the target gene. SHOX, a member of the bicoid subfamily of the paired homeobox family, controls fundamental aspects of growth and development and undergoes splicing resulting in two isoforms, SHOXA and SHOXB. SHOXA is expressed in skeletal muscle, placental, pancreas, heart and bone marrow fibroblast and, to a lesser extent, in kidney and lung. SHOXB is highly expressed in osteogenic cells, but not expressed in brain, kidney, liver or lung. Defects in the gene SHOXX cause Leri-Weill dyschondrosteosis (LWD) and Langer mesomelic dysplasia (LMD).
Specifications
| SHOX | |
| Polyclonal | |
| Unconjugated | |
| SHOX | |
| GCFX; growth control factor, X-linked; PHOG; Pseudoautosomal homeobox-containing osteogenic protein; short stature homeobox; short stature homeobox protein; Short stature homeobox-containing protein; SHOX; SHOXY; SS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6473 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15266 | |
| SHOX | |
| Recombinant Human SHOX. Recombinant protein control fragment (Product #RP-107034). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction