Learn More
SHOWA SHOWA S-TEX 541 Black Polyurethane Gloves
Product Code. 17737627
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Product Code. 17737627
Brand: SHOWA STEX54108L
Only null left
Add to basket
Add to basket
Get ultimate cut-resistant protection with SHOWA S-TEX 541 gloves. Stainless steel liner, Hagane Coil tech, and polyurethane coating provide top-notch defense against cuts and grime. Ergonomic design and seamless liner reduce hand fatigue and irritation.
  • The Showa S-Tex 541 is a revolutionary pair of gloves that provides superior protection against a variety of hazards while ensuring comfort and flexibility.
  • Designed for use in tough industrial and construction environments, these gloves are made with high-quality materials and advanced manufacturing techniques to ensure maximum durability and resistance to wear and tear.
  • The gloves are made with a high-performance polyethylene fiber that provides excellent resistance to cuts and slashes, making them ideal for use in industries where workers are exposed to sharp objects such as glass, metal, or blades. This makes them an excellent choice for construction workers, mechanics, and other professionals who work with machinery or sharp tools.
  • The S-Tex 541 also provides excellent protection against abrasions and punctures. The gloves are designed with a nitrile foam coating that provides a high level of grip and durability, even in wet or oily conditions. This coating also helps to prevent punctures and tears, making the gloves ideal for use in harsh environments.
  • The gloves are designed with an ergonomic shape that fits the natural curvature of the hand, reducing fatigue and increasing flexibility. The gloves are also breathable, allowing air to circulate around the hands and reducing the risk of sweating and discomfort.
  • The gloves are available in a range of sizes, from small to extra-large, ensuring that there is a pair of gloves to fit every hand. They are also designed to be easy to put on and take off, with an elasticated wrist that provides a secure and comfortable fit. This makes them ideal for use in fast-paced working environments, where workers need to be able to quickly and easily put on and remove their gloves.
Specifications
Coating Material Polyurethane
Color Black
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material Polyurethane, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN388:4X41D
Gender Unisex
Glove Cuff Style Knit