Learn More
SHOWA™ Series 8110 D-Flex™ Polyethylene and Spectra™ Fibre Gloves
Product Code. p-8010607
Click to view available options
Size:
X-Small
Medium
Large
X-Large
Unit Size:
Each
Product Code. 12940625
Brand: SHOWA™ 811009
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
SHOWA™ D-Flex Series 8110 Polyethylene and Spectra™ Fibre Gloves provide cut level 5 protection up to the wrist, and are flexible and durable
  • A flexible easy-maintenance glove providing effective protection against cuts
  • Durable, seamless knit from Spectra™ engineered fibre and high performance polyethylene (HPPE)
  • Uncoated with elasticated cuff and ambidextrous fit suitable for one-hand use
  • Standard finish without special treatment
  • Optimal dexterity and tactile feel retained
  • Lower allergy risks
  • No irritation by seams in contact with the skin
  • Great freedom of movement for the wrist
  • Fully launderable
  • Ambidextrous
  • Suitable for one-hand
Specifications
Certifications/Compliance EN388 2003, 2540, CE Cat II
Disposable Reusable
Hand Ambidextrous
Powdered Powder Free
Size Large
Sterility Non-sterile
Finger Style Full fingers
For Use With (Application) Glassware and food industries
Glove Cuff Style Elasticated
Length (Metric) 240 mm