missing translation for 'onlineSavingsMsg'
Learn More

SHOWA CS720 Blue Chemical Resistant Gloves

Product Code. 17817980
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
6 pairs
This item is not returnable. View return policy

Product Code. 17817980

Brand: SHOWA CS72010XL_OLD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The SHOWA CS720 chemical-resistant gloves offer a high level of protection against harmful substances, thanks to their double-dipped nitrile coating.

  • The gloves have a rough finish nitrile palm coating that gives a reliable grip.
  • The gloves come with a powdered liner that wicks away moisture, keeps wearers comfortable, and prevents slipping.
  • The CS720 gloves are durable and effective in preventing oil and dust from entering, allowing for optimal tactile precision.

Specifications

ANSI Abrasion Rating ANSI 1
Applications Industrial, Laboratory
Color Blue
Latex Free Yes
Material Nitrile, Polyester
Size X-Large
EN388 Standard EN388:3132X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLMNOPT
EN ISO 374 - 5 Bacteria, Fungi only
Lining Lined
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.