missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ N-DEX™ 7700PF Nighthawk Gloves

Product Code. 13151774
Change view
Click to view available options
Size:
Small
Large
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
13151774 Small -
11795409 Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13151774 Supplier SHOWA™ Supplier No. BST77001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Black, powder free static-dissipative gloves are ideal for a wide variety of industrial applications.

  • ESD properties
  • 100% chemical-resistant nitrile
  • Accelerator and powder free
  • Textured fingertips for enhanced tactility
  • Made in the U.S.A.
  • Ideal for a wide variety of applications including laboratory, food processing/handling, automotive, law enforcement and electronic assembly

Specifications

Certifications/Compliance FDA 21 CFR 170-199
Certifications/Compliance (Europe) Cat. III, CE Compliant 0123, EN 374-2, EN 374-3
Applications Industrial, Laboratory
AQL 1.5
Color Black
Disposable Single-use
Exterior Finish Partially Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model 7700PFTS
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.