Learn More
SHOWA 8110 Cut-Resistant Knit Gloves
Product Code. 17867980
Change view
Click to view available options
Size:
Small
Medium
Unit Size:
12 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17867980 Small 12 pairs
17877980 Medium 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17867980 Supplier SHOWA Supplier No. 811007S
  • ONLINE EXCLUSIVE PRICE
    £167.00
    £146.10 / 12 pairs
    Save £20.90
    12% Off
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The SHOWA 8110 gloves are a great choice for people who need strong and protective gloves for various industrial uses.
  • The gloves are designed to fit the hand naturally, reducing fatigue during prolonged use.
  • They offer optimal dexterity and breathability, keeping hands cool and comfortable even during intense physical activity.
  • The gloves are ideal for industries that require reliable cut protection, such as construction and manufacturing.
  • With exceptional durability, superior protection, and unparalleled comfort, the SHOWA 8110 gloves are the go-to choice for hand protection needs.
Specifications
Coating Material Uncoated
Color Gray
Hand Ambidextrous
Latex Free Yes
Material Glass Fiber, Polyethylene
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN388:2X43E
Gender Unisex
Glove Cuff Style Knit