missing translation for 'onlineSavingsMsg'
Learn More

SHOWA 690 Blue Lined Chemical Resistant Gloves with Extended Cuffs

Product Code. p-7244880
Click to view available options
Size:
Medium
Large
X-Large
Unit Size:
10 pairs

Product Code. 17831191

Brand: SHOWA 69008M

Only null left
Add to basket
Add to basket

The SHOWA 690 glove is designed for chemical protection and provides full coverage for the hand and arm, effectively shielding them from hazardous chemicals. The glove features extended bonded sleeves that are secured with an elasticated cuff.

  • Full coverage
  • Extended bonded sleeves
  • Features elasticated cuffs

Specifications

Chlorinated No
Applications Industrial, Laboratory
Color Blue
Latex Free Yes
Material Cotton, Polyester
Size Medium
EN388 Standard EN388:4121X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Extended
Lining Lined
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.