missing translation for 'onlineSavingsMsg'
Learn More

SHOWA 690 Blue Lined Chemical Resistant Gloves with Extended Cuffs

Product Code. 17851191
Change view
Click to view available options
Size:
Medium
Large
X-Large
Unit Size:
10 pairs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17851191 X-Large 10 pairs
17831191 Medium 10 pairs
17841191 Large 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 17851191 missing translation for 'mfr' SHOWA missing translation for 'supplierNo' 69010XL
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The SHOWA 690 glove is designed for chemical protection and provides full coverage for the hand and arm, effectively shielding them from hazardous chemicals. The glove features extended bonded sleeves that are secured with an elasticated cuff.

  • Full coverage
  • Extended bonded sleeves
  • Features elasticated cuffs

Specifications

Chlorinated No
Applications Industrial, Laboratory
Color Blue
Latex Free Yes
Material Cotton, Polyester
Size X-Large
EN388 Standard EN388:4121X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Extended
Lining Lined
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.