missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes - Phosphine

Product Code. p-4909574
Click to view available options
Components Measured:
Phosphine 0.01/a
Phosphine 0.1/C
Measurement Range:
0.01 to 1ppm
0.1 to 3ppm
Humidity Range:
40mg/L (max.)
Less than20mg/L
No. of Strokes:
1/3
3/10
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 10085891

Brand: Dräger™ 8101611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Description Short-Term Gas Detector Tubes
Components Measured Phosphine 0.01/a
Humidity Range Less than20mg/L
Measurement Range 0.01 to 1ppm
No. of Strokes 3/10
Quantity 10
Response Time 2.5 to 8 min.
Temperature Range (English) 35.6°F to 104°F
Temperature Range (Metric) 2° to 40°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.