missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Short Reducing Ferrules for 1/16 in. Fittings
15% Graphite / 85% Vespel ferrules provide leak-free system connections, leading to improved chromatography for non-MS systems
Brand: Trajan Scientific and Medical™ 073107
Code : 13
Additional Details : Weight : 0.50000kg
Description
- Composite of graphite and Vespel
- Mechanically robust, hard material, long lifetime
- Forms a strong grip with the capillary column
Specifications
1/16 in. Fittings | |
Graphite, Vespel | |
Reducing Ferrule | |
0.3 mm |
3.2 mm | |
2.7 mm | |
10/Pk. |