missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Shootin1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517167
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
SHTN1 is a protein coding gene involved in the generation of internal asymmetric signals required for neuronal polarization and neurite outgrowth. Mediates netrin-1-induced F-actin-substrate coupling or 'clutch engagement' within the axon growth cone through activation of CDC42, RAC1 and PAK1-dependent signaling pathway, thereby converting the F-actin retrograde flow into traction forces, concomitantly with filopodium extension and axon outgrowth. Plays a role in cytoskeletal organization by regulating the subcellular localization of phosphoinositide 3-kinase (PI3K) activity at the axonal growth cone. Plays also a role in regenerative neurite outgrowth. In the developing cortex, cooperates with KIF20B to promote both the transition from the multipolar to the bipolar stage and the radial migration of cortical neurons from the ventricular zone toward the superficial layer of the neocortex. involved in the accumulation of phosphatidylinositol 3,4,5-trisphosphate (PIP3) in the growth cone of primary hippocampal neurons.
Specifications
| Shootin1 | |
| Polyclonal | |
| Unconjugated | |
| SHTN1 | |
| 4930506M07Rik; Kiaa1598; mKIAA1598; RGD1311558; RIKEN cDNA 4930506M07 gene; shootin 1; Shootin1; shootin-1; shot1; Shtn1; similar to 4930506M07Rik protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 292139, 57698, 71653 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 30 μg/mL | |
| 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| A0MZ66, A0MZ67, Q8K2Q9 | |
| SHTN1 | |
| Synthetic peptide surrounding Lys450 of human shootin1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction