missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB30570.100uL
This item is not returnable.
View return policy
Description
Rabbit polyclonal antibody raised against partial recombinant human SHOC2.
- High specificity
- High reactivity
Specifications
| SHOC2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant human SHOC2. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| FLJ60412/KIAA0862/SIAA0862/SOC-2/SOC2/SUR-8/SUR8 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8036 | |
| Store at 4°C for short term storage. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunofluorescence (1 - 4 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500 - 1:1000) Western Blot (1:100 - 1:250) The optimal working dilution should be determined by the end user. | |
| Q9UQ13 | |
| SHOC2 | |
| Recombinant protein corresponding to amino acids 455-562 of human SHOC2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction