missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHMT2 Antibody (OTI3E9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74171CL1
This item is not returnable.
View return policy
Description
SHMT2 Monoclonal antibody specifically detects SHMT2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| SHMT2 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.1.2.1, GLY A+, GLYA, glycine auxotroph A, human complement for hamster, Glycine hydroxymethyltransferase, serine aldolase, serine hydroxymethylase, serine hydroxymethyltransferase 2 (mitochondrial), serine hydroxymethyltransferase, mitochondrial, Serine methylase, SHMT, threonine aldolase | |
| Full length human recombinant protein of human SHMT2(NP_005403) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| OTI3E9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 6472 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction