missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHMT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£255.00 - £402.00
Specifications
| Antigen | SHMT2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18446640
|
Novus Biologicals
NBP1-80754-25ul |
25ul |
£255.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18246888
|
Novus Biologicals
NBP1-80754 |
0.1 mL |
£402.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHMT2 Polyclonal specifically detects SHMT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SHMT2 | |
| Polyclonal | |
| Rabbit | |
| Rat, Human, Mouse | |
| P34897 | |
| 6472 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NTCPGDRSAITPGGLRLGAPALTSRQFREDDFRRVVDFIDEGVNIGLEVKSKTAKLQDFKSFLLKDSETSQRLANLRQRVEQFARAF | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 2.1.2.1, GLY A+, GLYA, glycine auxotroph A, human complement for hamster, Glycine hydroxymethyltransferase, serine aldolase, serine hydroxymethylase, serine hydroxymethyltransferase 2 (mitochondrial), serine hydroxymethyltransferase, mitochondrial, Serine methylase, SHMT, threonine aldolase | |
| SHMT2 | |
| IgG | |
| Affinity Purified | |
| Specificity of human SHMT2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title