missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SHMT1 Monoclonal Antibody (K56009_4G3), Invitrogen™

Mouse Monoclonal Antibody

£380.00 - £570.00

Specifications

Antigen SHMT1
Clone K56009_4G3
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30212329
View Documents
Invitrogen™
MA556123
200 μL
£570.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30210798
View Documents
Invitrogen™
MA555743
50 μL
£380.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence of this protein is as follows: MTMPVNGAHK DADLWSSHDK MLAQPLKDSD VEVYNIIKKE SNRQRVGLEL IASENFASRA VLEALGSCLN NKYSEGYPGQ RYYGGTEFID ELETLCQKRA LQAYKLDPQC WGVNVQPYSG SPANFAVYTA LVEPHGRIMG LDLPDGGHLT HGFMTDKKKI SATSIFFESM PYKVNPDTGY INYDQLEENA RLFHPKLIIA GTSCYSRNLE YARLRKIADE NGAYLMADMA HISGLVAAGV VPSPFEHCHV VTTTTHKTLR GCRAGMIFYR KGVKSVDPKT GKEILYNLES LINSAVFPGL QGGPHNHAIA GVAVALKQAM TLEFKVYQHQ VVANCRALSE ALTELGYKIV TGGSDNHLIL VDLRSKGTDG GRAEKVLEAC SIACNKNTCP GDRSALRPSG LRLGTPALTS RGLLEKDFQK VAHFIHRGIE LTLQIQSDTG VRATLKEFKE RLAGDKYQAA VQALREEVES FASLFPLPGL PDF

SHMT1 Monoclonal antibody specifically detects SHMT1 in Human samples. It is validated for Western Blot
Specifications

Specifications

SHMT1
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
AI324848; AI385541; C81125; CSHMT; cytoplasmic serine hydroxymethyltransferase; cytosolic form; cytosolic serine hydroxymethyltransferase; glycine hydroxymethyltransferase; LRRGT00032; MEL-32; MGC15229; MGC24556; mshmt; mshmt1; mshmt2; serine hydroxymethyl transferase; serine hydroxymethyltransferase; serine hydroxymethyltransferase 1; serine hydroxymethyltransferase 1 (soluble); serine hydroxymethyltransferase 1 (soluble) isoform 1; serine hydroxymethyltransferase 2 (mitochondrial); serine hydroxymethyltransferase, cytosolic; Serine methylase; SHMT; Shmt1; shmt1 protein; SHMT2; zgc:66171; zgc:77524; zSHMT-1
Recombinant human SHMT1 (immunogen range is 1-483).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shmt1
K56009_4G3
Western Blot
Unconjugated
Mouse
Human
P34896
6470
IgG1
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.