missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHMT1 Antibody (AT26E5), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59437B
This item is not returnable.
View return policy
Description
SHMT1 Monoclonal antibody specifically detects SHMT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| SHMT1 | |
| Monoclonal | |
| Biotin | |
| 14 kDa protein, CSHMT, cytoplasmic serine hydroxymethyltransferase, Glycine hydroxymethyltransferase, MGC15229, MGC24556, serine hydroxymethyltransferase 1 (soluble), serine hydroxymethyltransferase, cytosolic, Serine methylase, SHMTEC 2.1.2.1 | |
| Recombinant human SHMT1 (1-483aa) purified from E. coli | |
| 0.1 mL | |
| metabolism | |
| 6470 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| AT26E5 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction