missing translation for 'onlineSavingsMsg'
Learn More

SHMT1 Antibody (4F9), Novus Biologicals™

Product Code. 18376319 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376319 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376319 Supplier Novus Biologicals Supplier No. H00006470M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SHMT1 Monoclonal antibody specifically detects SHMT1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHMT1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 4F9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004160
Gene Alias 14 kDa protein, CSHMT, cytoplasmic serine hydroxymethyltransferase, Glycine hydroxymethyltransferase, MGC15229, MGC24556, serine hydroxymethyltransferase 1 (soluble), serine hydroxymethyltransferase, cytosolic, Serine methylase, SHMTEC 2.1.2.1
Host Species Mouse
Immunogen SHMT1 (NP_004160, 374 a.a. ~ 482 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EKVLEACSIACNKNTCPGDRSALRPSGLRLGTPALTSRGLLEKDFQKVAHFIHRGIELTLQIQSDTGVRATLKEFKERLAGDKYQAAVQALREEVESFASLFPLPGLPD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6470
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.