missing translation for 'onlineSavingsMsg'
Learn More

SHIP2/INPPL1 Antibody [CoraFluor™ 1], Novus Biologicals™

Product Code. 30732306 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30732306 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732306 Supplier Novus Biologicals Supplier No. AF5389CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

SHIP2/INPPL1 Polyclonal antibody specifically detects SHIP2/INPPL1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHIP2/INPPL1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias EC 3.1.3, EC 3.1.3.n1, inositol polyphosphate phosphatase-like 1, Inositol polyphosphate phosphatase-like protein 1, INPPL-1, phosphatidylinositol-34,5-trisphosphate 5-phosphatase 2, Protein 51C, SH2 domain-containing inositol phosphatase 2, SH2 domain-containing inositol-5'-phosphatase 2, SHIP-2, SHIP251C protein
Host Species Sheep
Immunogen E. coli-derived recombinant human SHIP2, Ala1106-Lys1258, Accession # O15357
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3636
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.