missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHIP Antibody (SHIP-01), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-81538JF549
This item is not returnable.
View return policy
Description
SHIP Monoclonal specifically detects SHIP in Human samples. It is validated for Flow Cytometry.
Specifications
| SHIP | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| INPP5D | |
| Peptide coresponding to a sequence within N-terminal domain of Human SHIP-1. | |
| 0.1 mL | |
| Primary | |
| The SHIP-01 reacts with SHIP-1, a phosphoinositide phosphatase largely confined to hematopoietic cells. Multiple forms of SHIP-1 have been reported with molecular weights of 110, 125, 130, 135 and 145 kDa. | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| SHIP-01 | |
| Flow Cytometry | |
| 145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, MGC104855, SHIPMGC142140 | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Immunology, Signal Transduction | |
| 3635 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction