missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHIP Antibody (SHIP-01) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-81538
This item is not returnable.
View return policy
Description
SHIP Monoclonal specifically detects SHIP in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| SHIP | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Flow Cytometry, Flow (Intracellular) 2-5 ug/ml, CyTOF-ready | |
| Q92835 | |
| INPP5D | |
| Peptide coresponding to a sequence within N-terminal domain of Human SHIP-1. | |
| Affinity Purified | |
| RUO | |
| 3635 | |
| Human | |
| IgG2a |
| Western Blot, Flow Cytometry, CyTOF | |
| SHIP-01 | |
| Unconjugated | |
| PBS (pH 7.4) with 15mM Sodium Azide | |
| 145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, MGC104855, SHIPMGC142140 | |
| Mouse | |
| 145 kDa | |
| 0.1 mg | |
| Primary | |
| The SHIP-01 reacts with SHIP-1, a phosphoinositide phosphatase largely confined to hematopoietic cells. Multiple forms of SHIP-1 have been reported with molecular weights of 110, 125, 130, 135 and 145 kDa. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction