missing translation for 'onlineSavingsMsg'
Learn More

Shikimic Acid, TRC

Product Code. 17964595
Change view
Click to view available options
Quantity:
50 mg
1 g
2.5 g
Unit Size:
1g
2.5g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17964595 1 g 1g
17941005 2.5 g 2.5g
17934705 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17964595 Supplier Toronto Research Chemicals Supplier No. TRCS3571001G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-59-0
Molecular Formula C7 H10 O5
Molecular Weight (g/mol) 174.15
Synonym 1-Cyclohexene-1-carboxylic acid, 3,4,5-trihydroxy-, (3R,4S,5R)- (9CI, ACI), (3R,4S,5R)-3,4,5-Trihydroxy-1-cyclohexene-1-carboxylic acid (ACI), 1-Cyclohexene-1-carboxylic acid, 3,4,5-trihydroxy-, [3R-(3α,4α,5β)]- (ZCI), (-)-3,4,5-Trihydroxy-1-cyclohexene-1-carboxylic acid, (-)-Shikimic acid, L-Shikimic acid, [3R-(3α,4α,5β)]-3,4,5-Trihydroxy-1-cyclohexene-1-carboxylic acid, NSC 59257, Shikimic acid
IUPAC Name (3R,4S,5R)-3,4,5-trihydroxycyclohexene-1-carboxylic acid
SMILES O[C@@H]1CC(=C[C@@H](O)[C@H]1O)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H10O5/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1,4-6,8-10H,2H2,(H,11,12)/t4-,5-,6-/m1/s1
Formula Weight 174.0528
Percent Purity >95%
Chemical Name or Material Shikimic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.