missing translation for 'onlineSavingsMsg'
Learn More

Shikimic acid, MedChemExpress

Product Code. 16428996
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428996 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428996 Supplier Medchem Express Supplier No. HYN0130
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Shikimic acid is a key metabolic intermediate of the aromatic amino acid biosynthesis pathway, found in microbes and plants.

Shikimic acid is a key metabolic intermediate of the aromatic amino acid biosynthesis pathway, found in microbes and plants.

Specifications

Chemical Name or Material Shikimic acid
CAS 138-59-0
Color White
Purity Grade Notes Research
Molecular Formula C7H10O5
Quantity 100 mg
Solubility Information H2O : 130 mg/mL (746.48 mM; Need ultrasonic)
SMILES O=C(C1=C[C@@H](O)[C@@H](O)[C@H](O)C1)O
Molecular Weight (g/mol) 174.15
Formula Weight 174.15
Percent Purity 99.15%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.