missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Shikimic Acid, 98%

Product Code. 10533491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10533491 1 g Glass bottle 1g
10697924 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10533491 Supplier Thermo Scientific Chemicals Supplier No. 132700010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 138-59-0 | C7H10O5 | 174.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-59-0
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
MDL Number MFCD00066278
InChI Key JXOHGGNKMLTUBP-HSUXUTPPSA-M
Synonym shikimic acid, shikimate, --shikimic acid, l-shikimic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-enecarboxylic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid, --shikimate, ccris 7681, unii-29ms2wi2nu, 3alpha,4alpha,5beta-trihydroxy-1-cyclohexene-1-carboxylic acid
PubChem CID 7057976
ChEBI CHEBI:36208
IUPAC Name (3R,4S,5R)-3,4,5-trihydroxycyclohexene-1-carboxylate
SMILES C1C(C(C(C=C1C(=O)[O-])O)O)O

Specifications

Melting Point 185.0°C to 191.0°C
Color Beige to White or Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Beilstein 10, 458
Merck Index 15, 8619
Specific Rotation Condition − 180.00 (25.00°C c=4,H2O)
Specific Rotation − 180.00
Solubility Information Solubility in water: 18g/100mL (20 c). Other solubilities: 2.25g/100mL abs.alcohol, 0.015g/100mL anhydr.ether, practically insoluble in chloroform, benzene and, petroleum ether
Formula Weight 174.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Shikimic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.