Learn More
Invitrogen™ Shigella Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA17245
Description
PA1-7245 detects Shigella sp. This antibody is polyvalent for Shigella antigens. The antiserum is unabsorbed and does cross-react with related Enterobacteriacea. PA1-7245 has been successfully used in ELISA and immunofluorescence procedures. The PA1-7245 immunogen is a mixture of Shigella boydii, Shigella flexneri and Shigella dysenteriae.
Specifications
| Shigella | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry | |
| 5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mixture of Shigella boydii, Shigella flexneri and Shigella dysenteriae. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.