missing translation for 'onlineSavingsMsg'
Learn More

Shigatoxin 1 Antibody (13C4), Novus Biologicals™

Product Code. 18346192 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346192 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346192 Supplier Novus Biologicals Supplier No. NBP311250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Shigatoxin 1 Monoclonal specifically detects Shigatoxin 1 in Bacteria samples. It is validated for Western Blot, Functional, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Shigatoxin 1
Applications Western Blot, Functional Assay, Immunoassay
Classification Monoclonal
Clone 13C4
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Functional, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Shiga toxin 1 subunit B, Shiga-like toxin 1, Stx1, STX1B, Verotoxin 1, VT-1
Host Species Mouse
Immunogen The monoclonal antibody 13C4 recognizes the 1B subunit of Shigatoxin 1.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 916677
Target Species Bacteria
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.