missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Shiga like-toxin 1 Monoclonal Antibody (13C4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA140277
This item is not returnable.
View return policy
Description
MA1-40277 detects Shigatoxin 1 from E. coli samples. MA1-40277 has been successfully used in ELISA, functional assays and Western blot applications. The MA1-40277 immunogen is 1B subunit of Shigatoxin 1.
Shiga-like toxins (SLTs), are also called Verotoxins. Enterohemorrhagic Escherichia coli (EHEC) strains which are primarily of serotypes 0157:H7, 026:H11 and O111:H8 have been incriminated as etiologic agents of hemorrhagic colitis and Hemolytic-uremic syndrome, a generalized disease characterized by acute renal failure, thrombocytopenia, and microangiopathic hemolytic anemia. There are several distinct E.coli SLTs. SLT-I and SLT-II are produced by EHEC. SLT-I and Shiga toxin share 99% deduced amino acid sequence homology, whereas SLT-I and SLT-II share about 60% deduced amino acid sequence homology. SLT-I and SLT-II are antigenically distinct. The protein structure of the toxin consists of two domains: the A polypeptide that inhibits protein synthesis by targeting ribosomes, and the B polypeptide pentamer that binds to the eukaryotic cell receptor globotriaosylceramide (Gb3) leading to receptor-mediated endocytosis.
Specifications
| Shiga like-toxin 1 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% BSA and no preservative | |
| Mouse | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA, Functional Assay, Western Blot | |
| 13C4 | |
| Unconjugated | |
| SLT1; SLT-1; SLTI; SLT-I; Verocytotoxin; Verotoxin | |
| 1B subunit of Shigatoxin 1. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction