missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHIELD Scientific SHIELDskin™ ORANGE NITRILE™ 260
Description
- Brand signature: Orange for biological risk
- Laboratory gloves for biohazard protection
- Latex gloves and nitrile/neoprene gloves
- Non-sterile and sterile gloves
- Maximum comfort
- Compliance with the latest European standards
- Category III PPE gloves (PPE Regulation (EU) 2016/425)
- Class 1 MDR (Regulation (EU) 2017/745)*
- Extra length from 260 to 300 mm (meets or exceeds ISO 21420:2020+A1:2022)
- Biohazard: ISO 374-5:2016
- Biosafety: AQL 0.25 (ISO 374-2:2019 Level 3)
- Virus resistant (ISO 16604:2004 Procedure B)
- Extensively tested for chemical permeation according to ISO 374-1:2016+A1:2018 (Test method EN 16523-1:2015+A1:2018)
- Reduced risk of allergies
- Double-walled protection afforded by the twinSHIELD™ technology
- Packaging designed to comply with laboratory environments processes
Specifications
Specifications
| Certifications/Compliance | AQL 0.25 |
| Certifications/Compliance (Europe) | PPER (EU) 2016/425, ISO 21420:2020, EN455-2:2015, ISO 374-1:2016-A1:2018 |
| Accelerator Free | Yes |
| Applications | Laboratory |
| AQL | 0.25 |
| Color | Orange |
| Disposable | Single-use |
| Exterior Finish | Fingertip Textured |
| Finger Thickness (English) | 6.7 mil |
| Finger Thickness (Metric) | 0.17 mm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?