missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHFM3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14013-25ul
This item is not returnable.
View return policy
Description
SHFM3 Polyclonal specifically detects SHFM3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SHFM3 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DAC, Dactylin, F-box and WD repeat domain containing 4, F-box and WD-40 domain protein 4, F-box and WD-40 domain-containing protein 4, F-box/WD repeat protein 4, F-box/WD repeat-containing protein 4, Fbw4, FBW4split hand/foot malformation (ectrodactyly) type 3, FBWD4, SHFM3, SHSF3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FBXW4 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: QCLHTIQTEDRVWSIAISPLLSSFVTGTACCGHFSPLRIWDLNSGQLMTHLGSDFPPGAGVLDVMYESPFTLLSCGYD | |
| 25ul | |
| Signal Transduction | |
| 6468 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction