missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHFM3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93850-0.1ml
This item is not returnable.
View return policy
Description
SHFM3 Polyclonal antibody specifically detects SHFM3 in Mouse samples. It is validated for Western Blot
Specifications
| SHFM3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DAC, Dactylin, F-box and WD repeat domain containing 4, F-box and WD-40 domain protein 4, F-box and WD-40 domain-containing protein 4, F-box/WD repeat protein 4, F-box/WD repeat-containing protein 4, Fbw4, FBW4split hand/foot malformation (ectrodactyly) type 3, FBWD4, SHFM3, SHSF3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 143-412 of human FBXW4 (NP_071322.1). QFRPDGASLNRRPLGVFAGHDEDVCHFVLANSHIVSAGGDGKIGIHKIHSTFTVKYSAHEQEVNCVDCKGGIIVSGSRDRTAKVWPLASGRLGQCLHTIQTEDRVWSIAISPLLSSFVTGTACCGHFSPLRIWDLNSGQLMTHLGSDFPPGAGVLDVMYESPFTLLSCGYDTYVRYWDLRTSVRKCVMEWEEPHDSTLYCLQTDGNHLLATGSSYYGVVRLWDRRQRACLHAFPLTSTPLSSPVYCLRLTTKHLYAALSYNLHVLDFQNP | |
| 0.1 mL | |
| Signal Transduction | |
| 6468 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction